Få mere at vide
rac-Parconazole Oxalate, TRC
Artikelnummer. 30137621
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137621 1 g
17991295 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137621 Leverandør Toronto Research Chemicals Leverandørnr. TRCP1932701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68685-55-2
Molekylær formel C17 H16 Cl2 N2 O3 . C2 H2 O4
Molekylvægt (g/mol) 457.261
Synonym 1-[[(2R,4S)-2-(2,4-Dichlorophenyl)-4-[(2-propyn-1-yloxy)methyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole Ethanedioate, cis-(±)-1-[[2-(2,4-Dichlorophenyl)-4-[(2-propynyloxy)methyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole, cis-1-[[2-(2,4-Dichlorophenyl)-4-[(2-propynyloxy)methyl]-1,3-dioxolan-2-yl]methyl]-1H-imidazole Ethanedioate;
IUPAC navn 1-[[(2R,4S)-2-(2,4-dichlorophenyl)-4-(prop-2-ynoxymethyl)-1,3-dioxolan-2-yl]methyl]imidazole;oxalic acid
SMIL OC(=O)C(=O)O.Clc1ccc(c(Cl)c1)[C@@]2(Cn3ccnc3)OC[C@H](COCC#C)O2
Tekniske data
Kemisk navn eller materiale rac-Parconazole Oxalate
Mængde 1 g
InChI formel InChI=1S/C17H16Cl2N2O3.C2H2O4/c1-2-7-22-9-14-10-23-17(24-14,11-21-6-5-20-12-21)15-4-3-13(18)8-16(15)19;3-1(4)2(5)6/h1,3-6,8,12,14H,7,9-11H2;(H,3,4)(H,5,6)/t14-,17-;/m0./s1
Formel vægt 456.049