Få mere at vide
rac-Monepantel Sulfone, TRC
Artikelnummer. 30010681
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010681 100 mg
17933455 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30010681 Leverandør Toronto Research Chemicals Leverandørnr. TRCM515810100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 851976-52-8
Molekylær formel C20H13F6N3O4S
Molekylvægt (g/mol) 505.39
Synonym Benzamide, N-[1-cyano-2-[5-cyano-2-(trifluoromethyl)phenoxy]-1-methylethyl]-4-[(trifluoromethyl)sulfonyl]-, N-[1-Cyano-2-[5-cyano-2-(trifluoromethyl)phenoxy]-1-methylethyl]-4-[(trifluoromethyl)sulfonyl]benzamide, Monepantel sulfone, N-(2-Cyano-1-(5-cyano-2-(trifluoromethyl)phenoxy)propan-2-yl)-4-((trifluoromethyl)sulfonyl)benzamide, Benzamide, N-[1-cyano-2-[5-cyano-2-(trifluoromethyl)phenoxy]-1-methylethyl]-4-[(trifluoromethyl)sulfonyl]-, N-[1-Cyano-2-[5-cyano-2-(trifluoromethyl)phenoxy]-1-methylethyl]-4-[(trifluoromethyl)sulfonyl]benzamide, Monepantel sulfone
IUPAC navn N-[2-cyano-1-[5-cyano-2-(trifluoromethyl)phenoxy]propan-2-yl]-4-(trifluoromethylsulfonyl)benzamide
SMIL CC(COc1cc(ccc1C(F)(F)F)C#N)(NC(=O)c2ccc(cc2)S(=O)(=O)C(F)(F)F)C#N
Tekniske data
Kemisk navn eller materiale rac-Monepantel Sulfone
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C20H13F6N3O4S/c1-18(10-28,11-33-16-8-12(9-27)2-7-15(16)19(21,22)23)29-17(30)13-3-5-14(6-4-13)34(31,32)20(24,25)26/h2-8H,11H2,1H3,(H,29,30)
Formel vægt 505.0531
Procent renhed >95%