Få mere at vide
rac Desethyl Oxybutynin-d5 Hydrochloride, TRC
Artikelnummer. 30353629
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353629 10 mg
30351799 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353629 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28947810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1173018-49-9
Molekylær formel C20H22D5NO3 . HCl
Molekylvægt (g/mol) 370.9
Synonym alpha-Cyclohexyl-alpha-hydroxybenzeneacetic Acid 4-[(Ethyl-d5)amino]-2-butynyl Ester Hydrochloride Salt
SMIL Cl.[2H]C([2H])([2H])C([2H])([2H])NCC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
Tekniske data
Kemisk navn eller materiale rac Desethyl Oxybutynin-d5 Hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C20H27NO3.ClH/c1-2-21-15-9-10-16-24-19(22)20(23,17-11-5-3-6-12-17)18-13-7-4-8-14-18;/h3,5-6,11-12,18,21,23H,2,4,7-8,13-16H2,1H3;1H/i1D3,2D2;
Formel vægt 370.2072