Få mere at vide
rac Darifenacin-d4, TRC
Artikelnummer. 30362212
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362212 10 mg
30360443 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362212 Leverandør Toronto Research Chemicals Leverandørnr. TRCD19340210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189701-43-6
Molekylær formel C28 2H4 H26 N2 O2
Molekylvægt (g/mol) 430.57
Synonym 2,2-Diphenyl-2-[1-[1,1,2,2-tetradeuterio-2-(2,3-dihydro-1-benzofuran-5-yl)ethyl]pyrrolidin-3-yl]acetamide, rac-Darifenacin D4 (ethyl D4), rac Darifenacin-d4
IUPAC navn 2,2-diphenyl-2-[1-[1,1,2,2-tetradeuterio-2-(2,3-dihydro-1-benzofuran-5-yl)ethyl]pyrrolidin-3-yl]acetamide
SMIL [2H]C([2H])(N1CCC(C1)C(C(=O)N)(c2ccccc2)c3ccccc3)C([2H])([2H])c4ccc5OCCc5c4
Tekniske data
Kemisk navn eller materiale rac Darifenacin-d4
Mængde 10 mg
InChI formel InChI=1S/C28H30N2O2/c29-27(31)28(23-7-3-1-4-8-23,24-9-5-2-6-10-24)25-14-17-30(20-25)16-13-21-11-12-26-22(19-21)15-18-32-26/h1-12,19,25H,13-18,20H2,(H2,29,31)/i13D2,16D2
Formel vægt 430.2558