missing translation for 'onlineSavingsMsg'
Få mere at vide

rac 1-Hydroxy Ketorolac Methyl Ester, TRC

Artikelnummer. 30008060
Change view
Click to view available options
Mængde:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30008060 25 mg -
30005177 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008060 Supplier Toronto Research Chemicals Supplier No. TRCH94365525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1391051-90-3
Molekylær formel C16H15NO4
Molekylvægt (g/mol) 285.29
Synonym 5-Benzoyl-1-hydroxy-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid Methyl Ester, Ketorolac Impurity G;
SMIL COC(=O)C1(CCN2C1=CC=C2C(=O)C3=CC=CC=C3)O

Specifications

Kemisk navn eller materiale rac 1-Hydroxy Ketorolac Methyl Ester
Mængde 25 mg
InChI formel InChI=1S/C16H15NO4/c1-21-15(19)16(20)9-10-17-12(7-8-13(16)17)14(18)11-5-3-2-4-6-11/h2-8,20H,9-10H2,1H3
Formel vægt 285.1001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.