Få mere at vide
rac 1-Hydroxy Ketorolac Methyl Ester, TRC
Artikelnummer. 30005177
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30005177 2.5 mg 2.5mg
30008060 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005177 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9436552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391051-90-3
Molekylær formel C16H15NO4
Molekylvægt (g/mol) 285.29
Synonym 5-Benzoyl-1-hydroxy-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid Methyl Ester, Ketorolac Impurity G;
SMIL COC(=O)C1(CCN2C1=CC=C2C(=O)C3=CC=CC=C3)O
Tekniske data
Kemisk navn eller materiale rac 1-Hydroxy Ketorolac Methyl Ester
Mængde 2.5 mg
InChI formel InChI=1S/C16H15NO4/c1-21-15(19)16(20)9-10-17-12(7-8-13(16)17)14(18)11-5-3-2-4-6-11/h2-8,20H,9-10H2,1H3
Formel vægt 285.1001