missing translation for 'onlineSavingsMsg'
Få mere at vide

R406, MedChemExpress

Artikelnummer. 16474968
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16474968 10 mg
16441217 50 mg
16437707 5 mg
16425746 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16474968 Leverandør Medchem Express Leverandørnr. HY12067

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress R406 is an orally available and competitive Syk/FLT3 inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. R406 reduces immune complex-mediated inflammation. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

R406 is an orally available and competitive Syk/FLT3 inhibitor for ATP binding with a Ki of 30 nM, potently inhibits Syk kinase activity in vitro with an IC50 of 41 nM, measured at an ATP concentration corresponding to its Km value. R406 reduces immune complex-mediated inflammation. R406 also inhibits Lyn (IC50=63 nM) and Lck (IC50=37 nM).

Tekniske data

Kemisk navn eller materiale R406
CAS 841290-81-1
Farve Earth Yellow
Renhedsgrad noter Research
Molekylær formel C28H29FN6O8S
Mængde 10 mg
Opløselighedsinformation DMSO : ≥ 61 mg/mL (97.04 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL COC1=CC(NC2=NC(NC3=NC(N4)=C(C=C3)OC(C)(C)C4=O)=C(C=N2)F)=CC(OC)=C1OC.OS(C5=CC=CC=C5)(=O)=O
Molekylvægt (g/mol) 628.63
Formel vægt 628.63
Procent renhed 96.67%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.