Få mere at vide
R1487 Hydrochloride, MedChemExpress
Artikelnummer. 16492397
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16492397 50 mg
16446278 10 mg
16443456 25 mg
16436616 100 mg
16418867 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16492397 Leverandør Medchem Express Leverandørnr. HY14975
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
MedChemExpress R1487 Hydrochloride is a highly potent and selective p38α inhibitor, with Kd values of 0.2 nM and 29 nM for p38α and p38β, respectively.
R1487 Hydrochloride is a highly potent and selective p38α inhibitor, with Kd values of 0.2 nM and 29 nM for p38α and p38β, respectively.
Tekniske data
Kemisk navn eller materiale R1487 Hydrochloride
CAS 449808-64-4
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C19H19ClF2N4O3
Mængde 50 mg
Opløselighedsinformation DMSO : 20.83 mg/mL (49.03 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMIL O=C1C(OC2=CC=C(F)C=C2F)=CC3=CN=C(NC4CCOCC4)N=C3N1C.Cl
Molekylvægt (g/mol) 424.83
Formel vægt 424.83
Procent renhed 98.68%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease