Få mere at vide
(R)-Sodium Danshensu, TRC
Artikelnummer. 30421839
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421839 5 mg
30409046 1 mg
30426685 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421839 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6219055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81075-52-7
Molekylær formel C9 H9 Na O5
Molekylvægt (g/mol) 220.155
Synonym Tanshinol Sodium, Salvianic Acid A Sodium, Dan Shen Suan A Sodium Salt, (alphaR)-alpha,3,4-Trihydroxy-benzenepropanoic Acid Monosodium Salt, (R)-alpha,3,4-Trihydroxy-benzenepropanoic Acid Monosodium Salt
IUPAC navn [(2R)-3-(3,4-dihydroxyphenyl)-2-hydroxypropanoyl]oxysodium
SMIL O[C@H](Cc1ccc(O)c(O)c1)C(=O)O[Na]
Tekniske data
Kemisk navn eller materiale (R)-Sodium Danshensu
Mængde 5 mg
InChI formel InChI=1S/C9H10O5.Na/c10-6-2-1-5(3-7(6)11)4-8(12)9(13)14;/h1-3,8,10-12H,4H2,(H,13,14);/q;+1/p-1/t8-;/m1./s1
Formel vægt 220.035