Få mere at vide
(R)-Norfluoxetine, TRC
Artikelnummer. 30136002
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136002 25 mg
30010673 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136002 Leverandør Toronto Research Chemicals Leverandørnr. TRCN68109025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 130194-43-3
Molekylær formel C16 H16 F3 N O
Molekylvægt (g/mol) 295.3
Synonym Benzenepropanamine, γ-[4-(trifluoromethyl)phenoxy]-, (γR)- (9CI, ACI), (γR)-γ-[4-(Trifluoromethyl)phenoxy]benzenepropanamine (ACI), Benzenepropanamine, γ-[4-(trifluoromethyl)phenoxy]-, (R)- (ZCI), (3R)-3-Phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine, (R)-3-(4-Trifluoromethylphenoxy)-3-phenylpropylamine, (R)-3-Phenyl-3-(4-trifluoromethyl-phenoxy)-propylamine, (R)-Norfluoxetine, 1-[(1R)-3-Amino-1-phenylpropoxy]-4-(trifluoromethyl)benzene
IUPAC navn (3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine
SMIL NCC[C@@H](Oc1ccc(cc1)C(F)(F)F)c2ccccc2
Tekniske data
Kemisk navn eller materiale (R)-Norfluoxetine
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C16H16F3NO/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12/h1-9,15H,10-11,20H2/t15-/m1/s1
Formel vægt 295.1184
Procent renhed >95%