Få mere at vide
(R)-Norfluoxetine-d5 (Phenyl-d5), TRC
Artikelnummer. 30421450
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421450 1 mg
30413567 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421450 Leverandør Toronto Research Chemicals Leverandørnr. TRCN6810921MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217648-64-0
Molekylær formel C16H11D5F3NO
Molekylvægt (g/mol) 300.33
Synonym (γR)-γ-[4-(Trifluoromethyl)phenoxy](benzene-d5)propanamine, (R)-3-(4-Trifluoromethylphenoxy)-3-(phenyl-d5)propylamine
SMIL NCC[C@@H](OC1=CC=C(C(F)(F)F)C=C1)C2=C([2H])C([2H])=C([2H])C([2H])=C2[2H]
Tekniske data
Kemisk navn eller materiale (R)-Norfluoxetine-d5 (Phenyl-d5)
Mængde 1 mg
InChI formel InChI=1S/C16H16F3NO/c17-16(18,19)13-6-8-14(9-7-13)21-15(10-11-20)12-4-2-1-3-5-12/h1-9,15H,10-11,20H2/t15-/m1/s1/i1D,2D,3D,4D,5D
Formel vægt 300.1498