Få mere at vide
R-(-)-N-Demethyl Deprenyl-d5, TRC
Artikelnummer. 30358369
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358369 1 mg
30350654 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358369 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2309221MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217977-04-2
Molekylær formel C12H10D5N
Molekylvægt (g/mol) 178.29
Synonym (αR)-α-Methyl-N-2-propyn-1-yl-(benzene-d5)ethanamine, (-)-Desmethylselegiline-d5, (-)-N-Desmethylselegiline-d5, Desmethylselegiline-d5, L-Desmethyldeprenyl-d5, L-Nordeprenyl-d5, N-Desmethylselegiline-d5, Norselegiline-d5
SMIL [2H]C1=C(C(=C(C(=C1[2H])[2H])C[C@@H](C)NCC#C)[2H])[2H]
Tekniske data
Kemisk navn eller materiale R-(-)-N-Demethyl Deprenyl-d5
Mængde 1 mg
InChI formel InChI=1S/C12H15N/c1-3-9-13-11(2)10-12-7-5-4-6-8-12/h1,4-8,11,13H,9-10H2,2H3/t11-/m1/s1/i4D,5D,6D,7D,8D