Få mere at vide
(R)-Licarbazepine Acetate, TRC
Artikelnummer. 30136134
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136134 50 mg
30008895 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136134 Leverandør Toronto Research Chemicals Leverandørnr. TRCL39769050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 186694-45-1
Molekylær formel C17H16N2O3
Molekylvægt (g/mol) 296.32
SMIL CC(=O)O[C@@H]1Cc2ccccc2N(C(=O)N)c3ccccc13
Tekniske data
Kemisk navn eller materiale (R)-Licarbazepine Acetate