Få mere at vide
(R)-Fluoxetine Hydrochloride, TRC
Artikelnummer. 30133815
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133815 25 mg -
30006884 2.5 mg 2.5mg
30008203 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133815 Leverandør Toronto Research Chemicals Leverandørnr. TRCF59709025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 114247-09-5
Molekylær formel C17 H18 F3 N O . Cl H
Molekylvægt (g/mol) 345.79
Synonym Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, hydrochloride (1:1), (γR)-, Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, hydrochloride, (R)-, Benzenepropanamine, N-methyl-γ-[4-(trifluoromethyl)phenoxy]-, hydrochloride, (γR)- (9CI), (-)-(R)-Fluoxetine hydrochloride, (-)-Fluoxetine hydrochloride, (R)-(-)-Fluoxetine hydrochloride, (R)-Fluoxetine hydrochloride, (R)-N-Methyl-3-(4-trifluoromethylphenoxy)-3-phenylpropylamine hydrochloride
IUPAC navn (3R)-N-methyl-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propan-1-amine;hydrochloride
SMIL Cl.CNCC[C@@H](Oc1ccc(cc1)C(F)(F)F)c2ccccc2
Tekniske data
Kemisk navn eller materiale (R)-Fluoxetine hydrochloride
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C17H18F3NO.ClH/c1-21-12-11-16(13-5-3-2-4-6-13)22-15-9-7-14(8-10-15)17(18,19)20;/h2-10,16,21H,11-12H2,1H3;1H/t16-;/m1./s1
Formel vægt 345.1107
Procent renhed >95%