Få mere at vide
(R)-(+)-Carvedilol, TRC
Artikelnummer. 30136687
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136687 50 mg
17903615 10 mg
17990575 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136687 Leverandør Toronto Research Chemicals Leverandørnr. TRCC18463050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 95093-99-5
Molekylær formel C24H26N2O4
Molekylvægt (g/mol) 406.47
Synonym 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, (2R)- (9CI), 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-, (R)-, (+)-Carvedilol, (R)-Carvedilol, R-(+)-Carvedilol, (R)-Carvedilol
IUPAC navn (2R)-1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethylamino]propan-2-ol
SMIL COc1ccccc1OCCNC[C@@H](O)COc2cccc3[nH]c4ccccc4c23
Tekniske data
Kemisk navn eller materiale (R)-(+)-Carvedilol
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3/t17-/m1/s1
Formel vægt 406.1893
Procent renhed >95%