Få mere at vide
(R)-2-Thioxothiazolidine-4-carboxylic Acid-13C3, TRC
Artikelnummer. 30005441
Skift visning
Mængde:
0.5 mg
1 mg
10 mg
Pakningsstørrelse:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30005441 0.5 mg 0.5mg
30009364 1 mg -
30008913 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005441 Leverandør Toronto Research Chemicals Leverandørnr. TRCT3950020.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2704345-00-4
Molekylær formel C13C3H5NO2S2
Molekylvægt (g/mol) 166.2
Synonym (R)-2-Thioxo-4-thiazolidinecarboxylic Acid-13C3, (R)-Thiazolidine-2-thione-4-carboxylic Acid-13C3, (R)-TTCA-13C3, (R)-TTCA-13C3 (Thiram Metabolite), Raphanusamic Acid-13C3
IUPAC navn (R)-2-thioxothiazolidine-4-carboxylic-4,5-13C2 acid-13C
SMIL S=C1N[13C@H]([13C](O)=O)[13CH2]S1
Tekniske data
Kemisk navn eller materiale (R)-2-Thioxothiazolidine-4-carboxylic Acid-13C3
Renhedsgrad noter HPLC
Mængde 0.5 mg
InChI formel InChI=1S/C4H5NO2S2/c6-3(7)2-1-9-4(8)5-2/h2H,1H2,(H,5,8)(H,6,7)/t2-/m0/s1/i1+1,2+1,3+1
Formel vægt 165.99
Procent renhed >95%