Få mere at vide
(R)-(-)-2-Methylglutaric Acid, TRC
Artikelnummer. 30483320
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483320 1 g
30473054 100 mg
30476560 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483320 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3116031G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1115-81-7
Molekylær formel C6H10O4
Molekylvægt (g/mol) 146.14
Synonym (2R)-2-Methylpentanedioic Acid, (-)-alpha-Methylglutaric Acid, (2R)-2-Methylglutaric Acid, (2R)-2-Methylpentanedioic Acid, (R)-2-Methylpentanedioic Acid
IUPAC navn (2R)-2-methylpentanedioic acid
SMIL C[C@H](CCC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale (R)-(-)-2-Methylglutaric Acid
Mængde 1 g
InChI formel InChI=1S/C6H10O4/c1-4(6(9)10)2-3-5(7)8/h4H,2-3H2,1H3,(H,7,8)(H,9,10)/t4-/m1/s1
Formel vægt 146.0579