CAS RN 1115-81-7
IUPAC Navn:
2-methylpentanedioic acid
Synonymer:
α-methylglutaric acid (5-L-glutamyl)-L-glutamate ion(2-) α-methylene glutarate α-methylene glutarate, ion(2-)
Molekylvægt (g/mol):
146.14
Molekylformel:
C6H10O4
InChi Key:
AQYCMVICBNBXNA-UHFFFAOYNA-N
SMILES:
CC(CCC(O)=O)C(O)=O
1
–
1
af
1
Resultater
1
(R)-(-)-2-Methylglutaric Acid, TRC
CAS: 1115-81-7 Molekylær formel: C6H10O4 Molekylvægt (g/mol): 146.14 Synonym: (2R)-2-Methylpentanedioic Acid,(-)-alpha-Methylglutaric Acid,(2R)-2-Methylglutaric Acid,(2R)-2-Methylpentanedioic Acid,(R)-2-Methylpentanedioic Acid IUPAC navn: (2R)-2-methylpentanedioic acid SMIL: C[C@H](CCC(=O)O)C(=O)O