Få mere at vide
(R)-2,2-Dimethylcyclopropanecarboxylic Acid, TRC
Artikelnummer. 30359732
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359732 500 mg
30350893 1 g
30352520 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30359732 Leverandør Toronto Research Chemicals Leverandørnr. TRCD286250500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 28624-52-4
Molekylær formel C6H10O2
Molekylvægt (g/mol) 114.14
Synonym (1R)-2,2-Dimethylcyclopropanecarboxylic Acid, (-)-2,2-Dimethylcyclopropanecarboxylic Acid
IUPAC navn (1R)-2,2-dimethylcyclopropane-1-carboxylic acid
SMIL CC1(C)C[C@H]1C(=O)O
Tekniske data
Kemisk navn eller materiale (R)-2,2-Dimethylcyclopropanecarboxylic Acid
Mængde 500 mg
InChI formel InChI=1S/C6H10O2/c1-6(2)3-4(6)5(7)8/h4H,3H2,1-2H3,(H,7,8)/t4-/m0/s1
Formel vægt 114.0681