Få mere at vide
Quinidine Sulfate (>85%), TRC
Artikelnummer. 30426224
Skift visning
Mængde:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426224 1 g
30416129 50 g
30417775 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426224 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ6850401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 747-45-5
Molekylær formel C20 H24 N2 O2 . H2 O4 S
Molekylvægt (g/mol) 422.495
Synonym (9S)-6'-Methoxy-cinchonan-9-ol Sulfate Dihydrate, Quinidine Sulfate, Quinidine Sulfate
IUPAC navn (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid
SMIL COc1ccc2nccc([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)c2c1.OS(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Quinidine Sulfate
Mængde 1 g
InChI formel InChI=1S/C20H24N2O2.H2O4S/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4)/t13-,14-,19+,20-;/m0./s1
Formel vægt 422.151