Learn More
Quickfit™ Recovery Still Head Adapter
Recover vapor after distillation. QuickFit™ glass recovery head provides an enclosed conduit between flasks and condensers.
Brand: Quickfit SH1/44
366.70 DKK valid until 2025-10-26
Use promo code "24136" to get your promotional price.
Description
Recover vapor after distillation. QuickFit™glass recovery head provides an enclosed conduit between flasks and condensers.
Provides a simple vapour conduit between flasks and distillation condensers
Specifications
Borosilicate Glass | |
29/32 (Flask/Condenser) | |
Recovery Still Head Adapter |
Promotions
-
Multiple Manufacturers
Focus on Agri-Science Testing
Use promotion code 24136 to get amazing savings.Promo Code: 24136Expires: 2025-10-26
New Offer Save Up to 87%