Learn More
Quickfit™ Receiver with Vented Bend
Receive distillate in a round bottomed flask. The QuickFit™ reciever is jointed with ventend bend to avoid the problems of closed systems.
Brand: Quickfit RA2/23
Description
Receive distillate in a round bottomed flask. The QuickFit™ reciever is jointed with ventend bend to avoid the problems of closed systems.
- Bend with vent
- With sidearm to provide vent to atmosphere
- Avoids the problems of closed systems
Specifications
Borosilicate Glass | |
4.7 in. | |
24/29 |
120 mm | |
Side arm | |
Receiver Adapter |