Learn More
Quickfit™ Receiver with Vented Bend
Receive distillate in a round bottomed flask. The QuickFit™ reciever is jointed with ventend bend to avoid the problems of closed systems.
Brand:  Quickfit RA2/11
 View more versions of this product
Product Code. 11332095
  • 331.00 DKK / Each
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 30-09-2025
Description
Receive distillate in a round bottomed flask. The QuickFit™ reciever is jointed with ventend bend to avoid the problems of closed systems.
  • Bend with vent
  • With sidearm to provide vent to atmosphere
  • Avoids the problems of closed systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
3.14 in.
14/23
80 mm
Side arm
Receiver Adapter
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Receiver with Vented Bend > Height: 80mm; Cone size: 14/23; Socket Size: 14/23