Learn More
Quickfit™ Plain Short Stem Adapter
Feed distillate from condensers into receiving vessels. QuickFit™ plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
Brand:  Quickfit RA1/11
 View more versions of this product
Product Code. 12445366
  • 240.00 DKK / Each
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 05-08-2025
Description
Feed distillate from condensers into receiving vessels. QuickFit™ plain short stem adapters come in a wide variety of lengths from short stems to very long ones.
  • With short or long stem
  • Useful for feeding distillate from condensers into receiving vessels
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
190 mm
14/23
7.4 in.
Feeding distillate from condensers into receiving vessels
Receiver Adapter
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Plain Short Stem Adapter > Length: 190mm; Socket size: 14/23