Learn More
Quickfit™ Plain Bend Delivery Receiver
Plain bend without vent
Brand:  Quickfit RA9/23
 View more versions of this product
Product Code. 15547950
  • 1555.00 DKK / Pack of 5
Only null left
Estimated Shipment: 07-10-2025
Description
Quickfit™ Plain Bend Delivery Receiver adapters are available in a range of cone sizes.
  • Plain bend without vent
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
4.3 in.
Receiver Adapter
110 mm
24/29
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Plain Bend Delivery Receiver > Cone Size: 24/29; Socket Size: 19/26; Height: 110mm