Learn More
Quickfit™ Medium Neck Flat Bottom Flask
Useful in reducing problems associated with splashing and frothing.
Brand: Quickfit FF1L/3M
2917.86 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Made of Borosilicate glass
- With longer neck than standard Quickfit flasks
Specifications
1000 mL | |
Borosilicate Glass | |
190 mm | |
Medium |
Product information
File size: 2.8 MB
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
Quickfit™ Medium Neck Flat Bottom Flask > Capacity: 1000mL
Spot an opportunity for improvement?Share a Content Correction