Learn More
Quickfit™ Medium Neck Flat Bottom Flask
Useful in reducing problems associated with splashing and frothing.
Brand:  Quickfit FF100/3M
 View more versions of this product
Product Code. 15547830
  • 1915.00 DKK / Pack of 10
Only null left
Estimated Shipment: 10-11-2025
Description
  • Made of Borosilicate glass
  • With longer neck than standard Quickfit flasks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
Borosilicate Glass
130 mm
Medium
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Medium Neck Flat Bottom Flask > Capacity: 100mL