Learn More
Quickfit™ Borosilicate Glass Iodine Flask
Complies with BS 2735
Brand:  Quickfit FIB500/4
 View more versions of this product
Product Code. 15507850
  • 6490.00 DKK / Pack of 10
Only null left
Estimated Shipment: 17-09-2025
Description
Complies with BS 2735
  • Complies with BS 2735
  • For the determination of iodine
  • Flask has ground socket and cup shaped top
  • Stopper has extended handle which projects well above the liquid seal
Note: Stopper is not included; sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BS 2735
500 mL
2
Iodine stopper: SIB 29
Top style: Cup shaped
Borosilicate Glass
Iodine Flask
29/32
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Iodine Flask > Capacity: 500mL