Learn More
Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar
Made from borosilicate glass. Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar is perfect for adding precise chemical or liquid that needs to go into a solution. You can combine the required reagents in precise quantities to make a solution.
Brand: Lenz Laborglasinstrumente™ 03026058
Description
- With hollow hexagonal stopper NS 29/32
- With scale
- With inscription label
- Material: DURAN™
Specifications
500 mL | |
Conical Ground Glass Joint | |
NS 29/32 | |
105 mm |
Borosilicate Glass | |
Conical | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.