Learn More
Quickfit™ Borosilicate Glass Dufton Fractionating Column
Efficient fractionating column designed with an an accurately fitted Dufton glass spiral. Quickfit™ Borosilicate Glass Dufton Fractionating Column can be disassembled for cleaning.
Brand: Quickfit™ FC2/23
This item is not returnable.
View return policy
Description
- Glass column with accurately fitted Dufton glass spiral
- Good efficiency - liquid spiralling down column meets ascending vapour
- Spiral can be removed for cleaning
Specifications
300mm | |
Dufton Fractionating Column | |
Borosilicate Glass | |
ISO 3585 |
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Dufton Fractionating Column > Length: 435mm
Spot an opportunity for improvement?Share a Content Correction