Få mere at vide
Quetiapine Sulfoxide-d8, TRC
Artikelnummer. 30425491
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425491 1 mg
30413742 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425491 Leverandør Toronto Research Chemicals Leverandørnr. TRCQ5100321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1330238-38-4
Molekylær formel C21H17D8N3O3S
Molekylvægt (g/mol) 407.56
Synonym 2-[2-[4-(5-Oxidodibenzo[b,f][1,4]thiazepin-11-yl)-1-(piperazinyl-d8)]ethoxy]ethanol
SMIL OCCOCCN(C([2H])([2H])C1([2H])[2H])C([2H])([2H])C([2H])([2H])N1C2=NC3=CC=CC=C3[S+]([O-])C4=C2C=CC=C4
Tekniske data
Kemisk navn eller materiale Quetiapine Sulfoxide-d8
Mængde 1 mg
InChI formel InChI=1S/C21H25N3O3S/c25-14-16-27-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)28(26)20-8-4-2-6-18(20)22-21/h1-8,25H,9-16H2/i9D2,10D2,11D2,12D2
Formel vægt 407.2119