missing translation for 'onlineSavingsMsg'
Få mere at vide

Pyr10, MedChemExpress

Artikelnummer. 16450228
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16450228 5 mg -
16417768 10 mg 10mg
16403737 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16450228 Leverandør Medchem Express Leverandørnr. HY19408

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Pyr10 is a pyrazole derivative and a selective TRP cation 3 (TRPC3) inhibitor. Pyr10 inhibits Ca2+ influx in carbachol-stimulated TRPC3-transfected HEK293 cells with an IC50 of 0.72 μM (IC50 of 13.08 μM for store operated Ca2+ entry in BRL-2H3 cells). Pyr10 has the ability to distinguish between receptor-operated TRPC3 and native stromal interaction molecule 1 (STIM1)/Orai1 channels.

Pyr10 is a pyrazole derivative and a selective TRP cation 3 (TRPC3) inhibitor. Pyr10 inhibits Ca2+ influx in carbachol-stimulated TRPC3-transfected HEK293 cells with an IC50 of 0.72 μM (IC50 of 13.08 μM for store operated Ca2+ entry in BRL-2H3 cells). Pyr10 has the ability to distinguish between receptor-operated TRPC3 and native stromal interaction molecule 1 (STIM1)/Orai1 channels.

Tekniske data

Kemisk navn eller materiale Pyr10
CAS 1315323-00-2
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C18H13F6N3O2S
Mængde 5 mg
Opløselighedsinformation DMSO : 100 mg/mL (222.53 mM; Need ultrasonic)
SMIL O=S(C1=CC=C(C)C=C1)(NC2=CC=C(N3N=C(C(F)(F)F)C=C3C(F)(F)F)C=C2)=O
Molekylvægt (g/mol) 449.37
Formel vægt 449.37
Procent renhed 97.52%
Grad Research
Fysisk form Solid
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.