Learn More
Fisherbrand™ PTFE T-Type Connector - Bayonet
With Bayonet fitting. Fisherbrand™ PTFE T-Type Connector - Bayonet contains an improved connector system.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01600kg
 View more versions of this product
Product Code. 10752983
  • 110.00 DKK / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
  • For use with rigid and semi rigid tubing - glass, FEP or PTFE
  • Screw lock holds from 10-1mmHg to 4 bar
Specifications
Specifications
PTFE T-Type Connector
Autoclavable
PTFE
Flexible tubing in PTFE, Viton, silicone rubber or similar materials
1 Each
16 mm
280°C
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
10 mm
51.5 mm
-200°C
33.5 mm
White