Learn More
Fisherbrand™ PTFE T-Type Connector - Bayonet
With Bayonet fitting. Fisherbrand™ PTFE T-Type Connector - Bayonet contains an improved connector system.
101.00 DKK - 110.00 DKK
Specifications
Product Type | PTFE T-Type Connector |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Autoclavable | Autoclavable |
Material | PTFE |
For Use With | Flexible tubing in PTFE, Viton, silicone rubber or similar materials |
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
10446022
|
Fisherbrand™
|
6 mm |
101.00 DKK
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
10427552
|
Fisherbrand™
|
4.5 mm |
101.00 DKK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
10531052
|
Fisherbrand™
|
8 mm |
101.00 DKK
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
10752983
|
Fisherbrand™
|
10 mm |
110.00 DKK
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Description
- For use with rigid and semi rigid tubing - glass, FEP or PTFE
- Screw lock holds from 10-1mmHg to 4 bar
Specifications
PTFE T-Type Connector | |
Autoclavable | |
Flexible tubing in PTFE, Viton, silicone rubber or similar materials | |
1 Each | |
280°C |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
PTFE | |
-200°C | |
White |