Learn More
VITLAB™ PTFE Oval Magnetic Stirring Bar
Brand:  VITLAB™ 311297
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 16583622
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Specifications
Specifications
Permanent magnet AlNiCo V core
PTFE
Magnetic Stir Bar
PTFE
Oval
10 mm
For preparation, separation and concentration
3/Pk.
20 mm