Learn More
VITLAB™ PTFE Oval Magnetic Stirring Bar
Brand:  VITLAB™ 311197
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16573622
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Specifications
Specifications
Permanent magnet AlNiCo V core
For preparation, separation and concentration
PTFE
Oval
6 mm
3/Pk.
15 mm