Learn More
VITLAB™ PTFE Cross Magnetic Stirring Bar
Brand:  VITLAB™ 316197
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10488422
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Permanent magnet AlNiCo V core
For preparation, separation and concentration
20mm
PTFE
Cross
20 x 20 x 8mm
8mm
1/Pk.
20 mm