Learn More
VITLAB™ PTFE Cross Magnetic Stirring Bar
34.35 DKK - 785.00 DKK
Specifications
Material | PTFE |
---|---|
Quantity | 1/Pk. |
Shape | Cross |
For Use With (Application) | For preparation, separation and concentration |
Includes | Permanent magnet AlNiCo V core |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
16563632
|
VITLAB™
316097 |
10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10254551
|
VITLAB™
316097 |
10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10346141
|
VITLAB™
316297 |
25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16583632
|
VITLAB™
316297 |
25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16573632
|
VITLAB™
316197 |
20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10488422
|
VITLAB™
316197 |
20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10418622
|
VITLAB™
316397 |
30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16593632
|
VITLAB™
316397 |
30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16503642
|
VITLAB™
316497 |
38 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10779251
|
VITLAB™
316497 |
38 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
PTFE | |
Cross | |
Permanent magnet AlNiCo V core |
1/Pk. | |
For preparation, separation and concentration |