Learn More
VITLAB™ PTFE Cross Magnetic Stirring Bar
Brand:  VITLAB™ 316097
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 10254551
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Permanent magnet AlNiCo V core
For preparation, separation and concentration
10mm
PTFE
Cross
10 x 10 x 5mm
5mm
1/Pk.
10 mm