Learn More
JG Finneran Associates™ PTFE/Butyl Rubber Preassembled Screw Caps
Brand: JG Finneran Associates™ 5394F-09G
Additional Details : Weight : 0.01000kg
Specifications
Green | |
Yes | |
9mm red septa | |
Screw Cap |
PTFE, Butyl Rubber | |
Designed to work in Agilent or other robotic arm autosampler | |
1000/Pk. |