Få mere at vide
PTC-124, TRC
Artikelnummer. 30428461
Skift visning
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428461 250 mg
30413455 1 g
30426129 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428461 Leverandør Toronto Research Chemicals Leverandørnr. TRCP839830250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 775304-57-9
Molekylær formel C15H9FN2O3
Molekylvægt (g/mol) 284.24
Synonym Ataluren PCT 124, 3-[5-(2-Fluorophenyl)-1,2,4-oxadiazol-3-yl]-benzoic Acid, PTC-124
IUPAC navn 3-[5-(2-fluorophenyl)-1,2,4-oxadiazol-3-yl]benzoic acid
SMIL OC(=O)c1cccc(c1)c2noc(n2)c3ccccc3F
Tekniske data
Kemisk navn eller materiale 3-[5-(2-Fluorophenyl)-1,2,4-oxadiazol-3-yl]benzoic Acid
Mængde 250 mg
InChI formel InChI=1S/C15H9FN2O3/c16-12-7-2-1-6-11(12)14-17-13(18-21-14)9-4-3-5-10(8-9)15(19)20/h1-8H,(H,19,20)
Formel vægt 284.0597