Få mere at vide
Prucalopride, MedChemExpress
Artikelnummer. 16496396
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16496396 100 mg
16495968 10 mg
16462117 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16496396 Leverandør Medchem Express Leverandørnr. HY14151
MedChemExpress Prucalopride (R093877) is a drug acting as a selective, high affinity 5-HT4 receptor agonist(pKi=8.6/8.1 for 5-HT4a/4b); >150-fold higher affinity for 5-HT4 receptors than for other receptors.
Prucalopride (R093877) is a drug acting as a selective, high affinity 5-HT4 receptor agonist(pKi=8.6/8.1 for 5-HT4a/4b)
Tekniske data
Kemisk navn eller materiale Prucalopride
CAS 179474-81-8
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C18H26ClN3O3
Mængde 100 mg
Opløselighedsinformation H2O : 50 mg/mL (135.92 mM; Need ultrasonic) ∣DMSO : ≥ 31 mg/mL (84.27 mM)
SMIL ClC1=C(N)C(CCO2)=C2C(C(NC3CCN(CCCOC)CC3)=O)=C1
Molekylvægt (g/mol) 367.87
Formel vægt 367.87
Procent renhed 99.83%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation