Få mere at vide
Propan-2-yl 2-Sulfanylacetate, TRC
Artikelnummer. 30011465
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011465 100 mg
30010887 500 mg
30005936 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011465 Leverandør Toronto Research Chemicals Leverandørnr. TRCB524470100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 7383-61-1
Molekylær formel C5 H10 O2 S
Molekylvægt (g/mol) 134.19
Synonym 1-Methylethyl 2-Mercaptoacetate, Isopropyl 2-Mercaptoacetate, Isopropyl Mercaptoacetate, Isopropyl Thioglycolate, Thioglycolic Acid Isopropyl Ester;
IUPAC navn isopropyl 2-mercaptoacetate
SMIL CC(C)OC(=O)CS
Tekniske data
Kemisk navn eller materiale Propan-2-yl 2-Sulfanylacetate
Mængde 100 mg
InChI formel InChI=1S/C5H10O2S/c1-4(2)7-5(6)3-8/h4,8H,3H2,1-2H3
Formel vægt 134.0402