missing translation for 'onlineSavingsMsg'
Learn More

Thermo Fisher Scientific (Asheville) LLC Thermoflex 1400 Recirculating Chiller T1 IPR (Internal Pressure Relief) SPCL FTG Air cooled

Cooling capacity up to 24,000 watts (81,948 btu/hr), is configurable to meet application demands. Quiet performance (<58 dBA) designed to remain quiet over the entire operating range of the chiller while maintaining temperature control accuracy.

Brand:  Thermo Fisher Scientific (Asheville) LLC 111163010000003


40661.27 DKK valid until 2024-05-31
Use promo code "21682" to get your promotional price.



Additional Details : Weight : 42.00000kg

Product Code. 16474310

  • 43050.58 DKK / Each
In Stock
to see stock.
Add to basket

Description

Description

Cooling capacity up to 24,000 watts (81,948 btu/hr), is configurable to meet application demands. Quiet performance (<58 dBA) designed to remain quiet over the entire operating range of the chiller while maintaining temperature control accuracy.

  • Thermoflex 1400 recirculating chiller
  • T1 IPR (Internal Pressure Relief)
  • SPCL FTG Air cooled
Specifications

Specifications

CE, CSA Semi S2
Air or Water-cooled
9.5 L/min.
5°C to 40°C
Analog & Digital (available)
Up to 1,400 watts (60 Hz) 1,170 watts (50 Hz)
T1 - Turbine Pump
240 V 50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers