Learn More
Plas Labs™ Glove Box Plas Labs Compact
Brand:  Plas Labs™ 830-CP-10/EXP
Additional Details : Weight : 62.00000kg
Product Code. 15102877
  • 93280.00 DKK / Each
Estimated Shipment: 12-07-2024
to see stock.
Description
Description
size A, inner dims: 710mm x 580mm x 740mm (w x d x h)