missing translation for 'onlineSavingsMsg'
Få mere at vide

Primaquine-d3 Diphosphate, TRC

Artikelnummer. 30415845
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415845 10 mg
30410008 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30415845 Leverandør Toronto Research Chemicals Leverandørnr. TRCP73350210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1318852-20-8
Molekylær formel C15H24D3N3O9P2
Molekylvægt (g/mol) 458.36
Synonym N4-(6-Methoxy-8-quinolinyl)-1,4-pentanediamine-d3 Phosphate, 8-[(4-Amino-1-methylbutyl)amino]-6-methoxyquinoline-d3 Phosphate, (+/-)-Primaquine-d3 Phosphate, 6-Methoxy-8-[4-amino-1- methylbutylamino]quinoline-d3 Phosphate, NSC 27296-d3, Neo-Quipenyl-d3 Phosphate, Primachin-d3 Phosphate, Primaquin-d8 Phosphate, SN 13272-d3, WR 2975-d3, dl-Primaquine-d3 Phosphate
SMIL NCCCC(C)NC1=C2C(C=CC=N2)=CC(OC([2H])([2H])[2H])=C1

Tekniske data

Kemisk navn eller materiale Primaquine-d3 Diphosphate
Mængde 10 mg
InChI formel InChI=1S/C15H21N3O/c1-11(5-3-7-16)18-14-10-13(19-2)9-12-6-4-8-17-15(12)14/h4,6,8-11,18H,3,5,7,16H2,1-2H3/i2D3
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.