missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Prexasertib, MedChemExpress
Beskrivelse
Prexasertib (LY2606368) is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor with a Ki of 0.9 nM and an IC50 of 50=8 nM) and RSK1 (IC50=9 nM). Prexasertib causes double-stranded DNA breakage and replication catastrophe resulting in apoptosis. Prexasertib shows potent anti-tumor activity.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Prexasertib |
| CAS | 1234015-52-1 |
| Farve | Light Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C18H19N7O2 |
| Mængde | 25 mg |
| Synonym | LY2606368 |
| Opløselighedsinformation | DMSO : 16.67 mg/mL (45.62 mM; Need ultrasonic) |
| SMIL | NCCCOC(C=CC=C1OC)=C1C2=CC(NC3=NC=C(C#N)N=C3)=NN2 |
| Molekylvægt (g/mol) | 365.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?