missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Praseomium solution for ICP 10g/L HNO3 matrix, Fisher Chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
958.00 DKK - 4510.00 DKK
Specifications
Analyte or Component Names | Re |
---|---|
Element | Re |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12996364
![]() |
Fisher Chemical
S 243 100 |
100 mL |
958.00 DKK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12906374
|
Fisher Chemical
S 243 250 |
250 mL | N/A | ||||||
12916374
|
Fisher Chemical
S 243 500 |
500 mL | N/A | ||||||
Specifications
Re | |
Re |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for ICP. Praseodymium (Pr) 10000mg/l in HNO3 5%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Praseomium solution for ICP 10g/L HNO3 matrix, Fisher Chemical™