Få mere at vide
Potassium 4-formylphenyltrifluoroborate, 97%, Thermo Scientific™
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
Beskrivelse
Denne Thermo Scientific mærkevare var oprindeligt en del af Alfa Aesar produktportefølje. Noget dokumentation og etiketoplysninger kan referere til det gamle mærke. Originalen Alfa Aesar produkt-/varekode eller SKU-reference er ikke ændret som en del af mærkeovergangen til Thermo Scientific .
Applications
It is employed in the synthesis of unsaturated organotrifluoroborates via Wittig and Horner-Wadsworth-Emmons Olefination.
Solubility
Soluble in water.
Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is employed in the synthesis of unsaturated organotrifluoroborates via Wittig and Horner-Wadsworth-Emmons Olefination.
Solubility
Soluble in water.
Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Potassium 4-formylphenyltrifluoroborate |
| Smeltepunkt | >300°C |
| Mængde | 1 g |
| Følsomhed | Air Sensitive |
| Opløselighedsinformation | Soluble in water. |
| Formel vægt | 212.02 |
| Procent renhed | 97% |