Få mere at vide
Potassium 2-Iodobenzenesulfonate (>85%), TRC
Artikelnummer. 30179523
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179523 5 g
30179400 1 g
30179412 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179523 Leverandør Toronto Research Chemicals Leverandørnr. TRCS6380035G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 649698-93-1
Molekylær formel C6 H5 I O3 S . K
Molekylvægt (g/mol) 323.17
Synonym 2-Iodobenzenesulfonic Acid Potassium Salt
IUPAC navn potassium;2-iodobenzenesulfonate
SMIL [K].O=S(=O)(O)C=1C=CC=CC1I
Tekniske data
Kemisk navn eller materiale Potassium 2-Iodobenzenesulfonate
Mængde 5 g
InChI formel InChI=1S/C6H5IO3S.K/c7-5-3-1-2-4-6(5)11(8,9)10;/h1-4H,(H,8,9,10);/q;+1/p-1
Formel vægt 321.8563